Mg(ii) meso-tetraphenylporphine ≥95%

Supplier: Ambeed
Warning

Synonyms: Magnesium(II) meso-tetraphenylporphine

A1329444-250MG A1329444-1G A1329444-5G
77596-790EA 67.65 USD
77596-790 77621-564 77621-566
Mg(ii) meso-tetraphenylporphine ≥95%
Mg(ii) meso-tetraphenylporphine

Precaución: for Research Use Only

Formula: C₄₄H₂₈MgN₄
MW: 637.02 g/mol
MDL Number: MFCD00144717
CAS Number: 14640-21-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR