N-(tert-Butyl)-3-((5-methyl-2-((4-(2-(pyrrolidin-1-yl)ethoxy)phenyl)amino)pyrimidin-4-yl)amino)benzenesulfonamide dihydrochloride hydrate ≥97%

Supplier: Ambeed

A1329297-250MG A1329297-100MG
77867-340EA 98 USD
77867-340 77867-339
N-(tert-Butyl)-3-((5-methyl-2-((4-(2-(pyrrolidin-1-yl)ethoxy)phenyl)amino)pyrimidin-4-yl)amino)benzenesulfonamide dihydrochloride hydrate ≥97%
N-(tert-Butyl)-3-((5-methyl-2-((4-(2-(pyrrolidin-1-yl)ethoxy)phenyl)amino)pyrimidin-4-yl)amino)benzenesulfonamide dihydrochloride hydrate

Precaución: for Research Use Only

Formula: C₂₇H₄₀Cl₂N₆O₄S
MDL Number: MFCD28137677
CAS Number: 1374744-69-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR