7-Methyl-5-(3-(piperazin-1-ylmethyl)-1,2,4-oxadiazol-5-yl)-2-(4-(trifluoromethoxy)benzyl)isoindolin-1-one methanesulfonate ≥98%

Supplier: Ambeed

A1329231-100MG A1329231-250MG
77867-337EA 419 USD
77867-337 77867-338
7-Methyl-5-(3-(piperazin-1-ylmethyl)-1,2,4-oxadiazol-5-yl)-2-(4-(trifluoromethoxy)benzyl)isoindolin-1-one methanesulfonate ≥98%
7-Methyl-5-(3-(piperazin-1-ylmethyl)-1,2,4-oxadiazol-5-yl)-2-(4-(trifluoromethoxy)benzyl)isoindolin-1-one methanesulfonate

Precaución: for Research Use Only

Formula: C₂₅H₂₈F₃N₅O₆S
MDL Number: MFCD32645412
CAS Number: 1314217-69-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR