3-((9-Amino-7-ethoxyacridin-3-yl)diazenyl)pyridine-2,6-diamine ≥98%

Supplier: Ambeed
Warning

A1329217-10MG A1329217-1MG A1329217-5MG
77596-780EA 53.3 USD
77596-780 77596-782 77596-784
3-((9-Amino-7-ethoxyacridin-3-yl)diazenyl)pyridine-2,6-diamine ≥98%
3-((9-Amino-7-ethoxyacridin-3-yl)diazenyl)pyridine-2,6-diamine

Precaución: for Research Use Only

Formula: C₂₀H₁₉N₇O
MDL Number: MFCD32701920
CAS Number: 681282-09-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR