DSPE-PEG-Mal, M.W. ~2000

Supplier: Ambeed
Warning

A1328973-100MG A1328973-250MG
77596-776EA 101.48 USD
77596-776 77596-778
DSPE-PEG-Mal, M.W. ~2000
DSPE-PEG-Mal

Precaución: for Research Use Only

MDL Number: MFCD03098977
CAS Number: 1190924-04-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR