B2 ≥97%

Supplier: Ambeed
Danger

A1328377-10MG A1328377-1MG A1328377-5MG
77596-766EA 39.98 USD
77596-766 77596-768 77596-770
B2 ≥97%
B2

Precaución: for Research Use Only

Formula: C₂₀H₁₇ClN₄O₃
MDL Number: MFCD02039810
CAS Number: 115687-05-3
UN: 2811
ADR: 6.1,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR