(11aS)-10,11,12,13-Tetrahydro-5-hydroxy-3,7-di-1-pyrenyl-diindeno[7,1-de:1',7'-fg][1,3,2]dioxaphosphocin 5-oxide ≥97%

Supplier: Ambeed
Warning

A1327705-100MG A1327705-250MG A1327705-1G
77596-752EA 277.78 USD
77596-752 77596-754 77621-538
(11aS)-10,11,12,13-Tetrahydro-5-hydroxy-3,7-di-1-pyrenyl-diindeno[7,1-de:1',7'-fg][1,3,2]dioxaphosphocin 5-oxide ≥97%
(11aS)-10,11,12,13-Tetrahydro-5-hydroxy-3,7-di-1-pyrenyl-diindeno[7,1-de:1',7'-fg][1,3,2]dioxaphosphocin 5-oxide

Precaución: for Research Use Only

Formula: C₄₉H₃₁O₄P
MW: 714.74 g/mol
MDL Number: MFCD31707581
CAS Number: 1643684-79-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR