(2S,3S)-3-Methylpent-4-en-2-ol ≥98%

Supplier: Ambeed
Danger

A1327371-1G A1327371-100MG
77621-536EA 115.36 USD
77621-536 77621-534
(2S,3S)-3-Methylpent-4-en-2-ol ≥98%
(2S,3S)-3-Methylpent-4-en-2-ol

Precaución: for Research Use Only

Formula: C₆H₁₂O
MW: 100.1589 g/mol
MDL Number: MFCD32662356
CAS Number: 99438-31-0
UN: 1987
ADR: 3,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR