2-(Ethylcarbamoyl)pyridine-5-boronic acid pinacol ester ≥98%

Supplier: Ambeed
Warning

Synonyms: N-Ethyl-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)picolinamide

A1327056-1G A1327056-5G
77867-294EA 266 USD
77867-294 77867-295
2-(Ethylcarbamoyl)pyridine-5-boronic acid pinacol ester ≥98%
2-(Ethylcarbamoyl)pyridine-5-boronic acid pinacol ester

Precaución: for Research Use Only

Formula: C₁₄H₂₁BN₂O₃
MDL Number: MFCD13191367
CAS Number: 1006876-28-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR