(R)-4-(2-((5-Fluoro-6-methoxypyridin-3-yl)amino)-5-(1-(4-(methylsulfonyl)piperazin-1-yl)ethyl)pyridin-3-yl)-6-methyl-1,3,5-triazin-2-amine ≥98%

Supplier: Ambeed
Warning

Synonyms: AMG 511

A1326767-10MG A1326767-1MG A1326767-5MG
77621-524EA 350.2 USD
77621-524 77621-526 77621-528
(R)-4-(2-((5-Fluoro-6-methoxypyridin-3-yl)amino)-5-(1-(4-(methylsulfonyl)piperazin-1-yl)ethyl)pyridin-3-yl)-6-methyl-1,3,5-triazin-2-amine ≥98%
(R)-4-(2-((5-Fluoro-6-methoxypyridin-3-yl)amino)-5-(1-(4-(methylsulfonyl)piperazin-1-yl)ethyl)pyridin-3-yl)-6-methyl-1,3,5-triazin-2-amine

Precaución: for Research Use Only

Formula: C₂₂H₂₈FN₉O₃S
MW: 517.58 g/mol
MDL Number: MFCD25976575
CAS Number: 1253573-53-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR