1,2-Bis((2S,5S)-2,5-diethylphospholano)ethane(cyclooctadiene)rhodium(I) trifluoromethanesulfonate ≥98%
Supplier: Ambeed
|
Warning
|
A1326493-100MG
A1326493-250MG
A1326493-50MG
77621-518EA
160.68
USD
77621-518
77621-520
77621-522
1,2-Bis((2S,5S)-2,5-diethylphospholano)ethane(cyclooctadiene)rhodium(I) trifluoromethanesulfonate ≥98%
1,2-Bis((2S,5S)-2,5-diethylphospholano)ethane(cyclooctadiene)rhodium(I) trifluoromethanesulfonate
Precaución: for Research Use Only
Formula:
C₂₇H₄₈F₃O₃P₂RhS MW: 674.5815 g/mol |
CAS Number:
2709040-48-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...