1-(4-Chloro-3-(trifluoromethyl)phenyl)-3-(4-(4-cyanophenoxy)phenyl)urea 98+%

Supplier: Ambeed

A1326050-100MG A1326050-250MG
77867-283EA 296 USD
77867-283 77867-284
1-(4-Chloro-3-(trifluoromethyl)phenyl)-3-(4-(4-cyanophenoxy)phenyl)urea 98+%
1-(4-Chloro-3-(trifluoromethyl)phenyl)-3-(4-(4-cyanophenoxy)phenyl)urea

Precaución: for Research Use Only

Formula: C₂₁H₁₃ClF₃N₃O₂
MDL Number: MFCD27665015
CAS Number: 1313019-65-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR