4-Methyl-1H-indazol-5-amine ≥98%

Supplier: Ambeed
Danger

A132571-100MG A132571-250MG
77876-943EA 11 USD
77876-943 77876-944
4-Methyl-1H-indazol-5-amine ≥98%
4-Methyl-1H-indazol-5-amine

Precaución: for Research Use Only

Formula: C₈H₉N₃
MDL Number: MFCD09263208
CAS Number: 101257-89-0
UN: 2811
ADR: 6.1,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR