2-(Diphenylphosphaneyl)-N-((R)-(6-methoxyquinolin-4-yl)((1S,2R,4S,5R)-5-vinylquinuclidin-2-yl)methyl)benzamide ≥98%

Supplier: Ambeed
Warning

A1325605-100MG A1325605-250MG
77592-528EA 182.45 USD
77592-528 77621-506
2-(Diphenylphosphaneyl)-N-((R)-(6-methoxyquinolin-4-yl)((1S,2R,4S,5R)-5-vinylquinuclidin-2-yl)methyl)benzamide ≥98%
2-(Diphenylphosphaneyl)-N-((R)-(6-methoxyquinolin-4-yl)((1S,2R,4S,5R)-5-vinylquinuclidin-2-yl)methyl)benzamide

Precaución: for Research Use Only

Formula: C₃₉H₃₈N₃O₂P
MW: 611.7117 g/mol
CAS Number: 2271134-66-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR