(5-(Methoxycarbonyl)-1-tosyl-1H-pyrrol-3-yl)boronic acid ≥97%

Supplier: Ambeed
Warning

A132520-250MG A132520-100MG A132520-1G
77592-524EA 76.88 USD
77592-524 77683-173 77683-174
(5-(Methoxycarbonyl)-1-tosyl-1H-pyrrol-3-yl)boronic acid ≥97%
(5-(Methoxycarbonyl)-1-tosyl-1H-pyrrol-3-yl)boronic acid

Precaución: For research use only.

Formula: C₁₃H₁₄BNO₆S
MW: 323.12 g/mol
MDL Number: MFCD18427599
CAS Number: 916177-00-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR