3-Methoxy-5-(methoxycarbonyl)phenylboronic acid pinacol ester ≥98%

Supplier: Ambeed
Warning

Synonyms: Methyl 3-methoxy-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzoate, 3-Methoxy-5-(methoxycarbonyl)phenylboronic acid, pinacol ester

A132513-100MG A132513-1G A132513-250MG A132513-5G
77883-066EA 60 USD
77883-066 77883-067 77883-068 77883-069
3-Methoxy-5-(methoxycarbonyl)phenylboronic acid pinacol ester ≥98%
3-Methoxy-5-(methoxycarbonyl)phenylboronic acid pinacol ester

Precaución: for Research Use Only

Formula: C₁₅H₂₁BO₅
MW: 292.135 g/mol
MDL Number: MFCD12026074
CAS Number: 889654-06-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR