tert-Butyl 2-bromo-7-azaspiro[3.5]nonane-7-carboxylate ≥95%

Supplier: Ambeed
Warning

A1324987-100MG A1324987-250MG A1324987-1G A1324987-5G
77596-736EA 62.53 USD
77596-736 77596-738 77621-500 77621-502
tert-Butyl 2-bromo-7-azaspiro[3.5]nonane-7-carboxylate ≥95%
tert-Butyl 2-bromo-7-azaspiro[3.5]nonane-7-carboxylate

Precaución: for Research Use Only

Formula: C₁₃H₂₂BrNO₂
MW: 304.2233 g/mol
MDL Number: MFCD24471158
CAS Number: 1225276-07-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR