(E)-(3-Methylstyryl)boronic acid ≥97%

Supplier: Ambeed

A1324444-250MG A1324444-5G A1324444-100MG A1324444-10G A1324444-1G
77867-270EA 91 USD
77867-270 77867-271 77867-267 77867-268 77867-269
(E)-(3-Methylstyryl)boronic acid ≥97%
(E)-(3-Methylstyryl)boronic acid

Precaución: for Research Use Only

Formula: C₉H₁₁BO₂
MDL Number: MFCD06202538
CAS Number: 352530-26-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR