2'-Bromospiro[cyclopentane-1,9'-fluorene] ≥97%

Supplier: Ambeed
Warning

A1324284-1G A1324284-250MG
77621-496EA 29.87 USD
77621-496 77867-264
2'-Bromospiro[cyclopentane-1,9'-fluorene] ≥97%
2'-Bromospiro[cyclopentane-1,9'-fluorene]

Precaución: for Research Use Only

Formula: C₁₇H₁₅Br
MDL Number: MFCD28956164
CAS Number: 797056-47-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR