(4-Chloro-2-nitrophenyl)boronic acid ≥98%

Supplier: Ambeed

A1323494-100MG A1323494-250MG
77867-257EA 168 USD
77867-257 77867-258
(4-Chloro-2-nitrophenyl)boronic acid ≥98%
(4-Chloro-2-nitrophenyl)boronic acid

Precaución: for Research Use Only

Formula: C₆H₅BClNO₄
MDL Number: MFCD16295411
CAS Number: 1312921-23-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR