3-Propylidenephthalide ≥96% (mixture of cis and trans)

Supplier: Ambeed
Warning

A132341-1G A132341-5G
77878-951EA 5 USD
77878-951 77878-952
3-Propylidenephthalide ≥96% (mixture of cis and trans)
3-Propylidenephthalide

Precaución: for Research Use Only

Formula: C₁₁H₁₀O₂
MDL Number: MFCD00051813
CAS Number: 17369-59-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR