(2R,4S)-4-Amino-1-(tert-butoxycarbonyl)pyrrolidine-2-carboxylic acid ≥95%

Supplier: Ambeed
Warning

Synonyms: (4S)-1-Boc-4-amino-D-proline

A132178-100MG A132178-1G A132178-250MG
77670-868EA 33 USD
77670-868 77670-870 77670-872
(2R,4S)-4-Amino-1-(tert-butoxycarbonyl)pyrrolidine-2-carboxylic acid ≥95%
(2R,4S)-4-Amino-1-(tert-butoxycarbonyl)pyrrolidine-2-carboxylic acid

Precaución: for Research Use Only

Formula: C₁₀H₁₈N₂O₄
MW: 230.26 g/mol
MDL Number: MFCD08704543
CAS Number: 132622-78-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR