(2S,4S)-2,4-Dimethylazetidine hydrochloride ≥97%

Supplier: Ambeed
Warning

A131842-100MG A131842-250MG
77747-939EA 242 USD
77747-939 77747-940
(2S,4S)-2,4-Dimethylazetidine hydrochloride ≥97%
(2S,4S)-2,4-Dimethylazetidine hydrochloride

Precaución: for Research Use Only

Formula: C₅H₁₂ClN
MW: 121.61 g/mol
MDL Number: MFCD31705783
CAS Number: 470666-35-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR