2-Fluorobenzamidine hydrochloride ≥97%

Supplier: Ambeed
Warning

A131820-10G A131820-1G A131820-5G
77611-166EA 182.31 USD
77611-166 77891-923 77891-924
2-Fluorobenzamidine hydrochloride ≥97%
2-Fluorobenzamidine hydrochloride

Precaución: for Research Use Only

Formula: C₇H₈ClFN₂
MDL Number: MFCD04114435
CAS Number: 57075-81-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR