Roxithromycin ≥95%

Supplier: Ambeed
Warning

Synonyms: (3R,4S,5S,6R,7R,9R,11S,12R,13S,14R,E)-6-(((2S,3R,4S,6R)-4-(Dimethylamino)-3-hydroxy-6-methyltetrahydro-2H-pyran-2-yl)oxy)-14-ethyl-7,12,13-trihydroxy-4-(((2R,4R,5S,6S)-5-hydroxy-4-methoxy-4,6-dimethyltetrahydro-2H-pyran-2-yl)oxy)-10-(((2-methoxyethoxy)methoxy)imino)-3,5,7,9,11,13-hexamethyloxacyclotetradecan-2-one

A131067-100G A131067-1MG A131067-1G A131067-5G
77666-878EA 210 USD
77666-878 77666-880 77871-609 77871-610
Roxithromycin ≥95%
Roxithromycin

Precaución: for Research Use Only

Formula: C₄₁H₇₆N₂O₁₅
MDL Number: MFCD00214389
CAS Number: 80214-83-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR