cis-1,4-Cyclohexanedimethanol ≥95%

Supplier: Ambeed

A1308679-1G A1308679-250MG
77867-247EA 592 USD
77867-247 77867-248
cis-1,4-Cyclohexanedimethanol ≥95%
cis-1,4-Cyclohexanedimethanol

Precaución: for Research Use Only

Formula: C₈H₁₆O₂
MW: 144.21 g/mol
MDL Number: MFCD00064957
CAS Number: 3236-47-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR