Zinc(II) oleate ≥98%

Supplier: Ambeed

A1307655-25G A1307655-100G
77596-722EA 15.38 USD
77596-722 77867-246
Zinc(II) oleate ≥98%
Zinc(II) oleate

Precaución: for Research Use Only

Formula: C₃₆H₆₆O₄Zn
MW: 628.32 g/mol
MDL Number: MFCD00152954
CAS Number: 557-07-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR