WZ-8040 ≥98%

Supplier: Ambeed
Warning

A130565-10MG A130565-5MG
77685-134EA 140 USD
77685-134 77685-135
WZ-8040 ≥98%
WZ-8040

Precaución: for Research Use Only

Formula: C₂₄H₂₅ClN₆OS
MDL Number: MFCD18074511
CAS Number: 1214265-57-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR