Zinc dibenzoate ≥98%, Analytical reagent (AR)

Supplier: Ambeed
Danger

Synonyms: Benzoic acid, zinc salt (2:1), Benzoic acid zinc (II) salt

A1303238-1G A1303238-100G A1303238-25G A1303238-5G A1303238-500G
77592-506EA 9.23 USD
77592-506 77596-714 77596-716 77621-486 77867-239
Zinc dibenzoate ≥98%, Analytical reagent (AR)
Zinc dibenzoate

Precaución: for Research Use Only

Formula: C₁₄H₁₀O₄Zn
MDL Number: MFCD00050788
CAS Number: 553-72-0
UN: 3077
ADR: 9,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR