Heptyl-β-D-glucopyranoside ≥99%

Supplier: Ambeed
Warning

Synonyms: n-Heptyl β-D-glucopyranoside, (2R,3R,4S,5S,6R)-2-(Heptyloxy)-6-(hydroxymethyl)tetrahydro-2H-pyran-3,4,5-triol

A130294-100MG A130294-250MG
77685-131EA 37 USD
77685-131 77685-132
Heptyl-β-D-glucopyranoside ≥99%
Heptyl-β-D-glucopyranoside

Precaución: for Research Use Only

Formula: C₁₃H₂₆O₆
MW: 278.35 g/mol
MDL Number: MFCD00063299
CAS Number: 78617-12-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR