3,5-Di-tert-butyl-[1,1'-biphenyl]-4-ol ≥95%

Supplier: Ambeed
Danger

A1301859-1G A1301859-100MG A1301859-250MG A1301859-5G
77592-502EA 834.35 USD
77592-502 77621-480 77621-482 77621-484
3,5-Di-tert-butyl-[1,1'-biphenyl]-4-ol ≥95%
3,5-Di-tert-butyl-[1,1'-biphenyl]-4-ol

Precaución: for Research Use Only

Formula: C₂₀H₂₆O
MW: 282.42 g/mol
MDL Number: MFCD00082900
CAS Number: 2668-47-5
UN: 3077
ADR: 9 Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR