1,6-Dimethyl-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-indazole ≥95%

Supplier: Ambeed

A130177-100MG A130177-250MG
77783-334EA 34 USD
77783-334 77783-335
1,6-Dimethyl-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-indazole ≥95%
1,6-Dimethyl-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-indazole

Precaución: for Research Use Only

Formula: C₁₅H₂₁BN₂O₂
MDL Number: MFCD28718055
CAS Number: 1463055-05-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR