2-Oxo-3-piperidinecarboxylic acid ≥98%

Supplier: Ambeed
Warning

A129586-1G A129586-250MG A129586-100MG
77670-832EA 78 USD
77670-832 77670-834 77881-033
2-Oxo-3-piperidinecarboxylic acid ≥98%
2-Oxo-3-piperidinecarboxylic acid

Precaución: for Research Use Only

Formula: C₆H₉NO₃
MDL Number: MFCD08061931
CAS Number: 41888-21-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR