[1,2,4]Triazol-1-ylacetic acid ≥98%

Supplier: Ambeed
Warning

A129020-250MG A129020-1G
77615-008EA 14.42 USD
77615-008 77745-931
[1,2,4]Triazol-1-ylacetic acid ≥98%
[1,2,4]Triazol-1-ylacetic acid

Precaución: for Research Use Only

Formula: C₄H₅N₃O₂
MDL Number: MFCD00036051
CAS Number: 28711-29-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR