N-Carbobenzoxy-S-benzyl-L-cysteine ≥97%

Supplier: Ambeed
Warning

Synonyms: Cbz-Cys(Bzl)-OH, N-Cbz-S-benzyl-L-cysteine, Z-Cys(Bzl)-OH

A128740-25G A128740-5G A128740-1G
77871-594EA 68 USD
77871-594 77871-595 77871-593
N-Carbobenzoxy-S-benzyl-L-cysteine ≥97%
N-Carbobenzoxy-S-benzyl-L-cysteine

Precaución: for Research Use Only

Formula: C₁₈H₁₉NO₄S
MDL Number: MFCD00004785
CAS Number: 3257-18-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR