5,6-Diphenyl-3-(2-pyridyl)-1,2,4-triazine ≥98%
Supplier: Ambeed
|
Warning
|
A128167-1G
A128167-5G
77747-914EA
21
USD
77747-914
77747-915
5,6-Diphenyl-3-(2-pyridyl)-1,2,4-triazine ≥98%
5,6-Diphenyl-3-(2-pyridyl)-1,2,4-triazine
Precaución: for Research Use Only
Formula:
C₂₀H₁₄N₄ |
MDL Number:
MFCD00006462 CAS Number: 1046-56-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...