(S)-2-(2-((S)-(4-(tert-Butyl)phenyl)sulfinyl)phenyl)-4-phenyl-4,5-dihydrooxazole ≥98%

Supplier: Ambeed
Warning

A1281591-100MG A1281591-250MG A1281591-1G
77621-472EA 135.96 USD
77621-472 77621-474 77867-230
(S)-2-(2-((S)-(4-(tert-Butyl)phenyl)sulfinyl)phenyl)-4-phenyl-4,5-dihydrooxazole ≥98%
(S)-2-(2-((S)-(4-(tert-Butyl)phenyl)sulfinyl)phenyl)-4-phenyl-4,5-dihydrooxazole

Precaución: for Research Use Only

Formula: C₂₅H₂₅NO₂S
CAS Number: 2242702-44-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR