(S)-2-(2-((R)-(4-(tert-Butyl)phenyl)sulfinyl)phenyl)-4-phenyl-4,5-dihydrooxazole ≥95%

Supplier: Ambeed

A1281569-100MG A1281569-1G A1281569-250MG A1281569-5G
77867-225EA 145 USD
77867-225 77867-226 77867-227 77867-228
(S)-2-(2-((R)-(4-(tert-Butyl)phenyl)sulfinyl)phenyl)-4-phenyl-4,5-dihydrooxazole ≥95%
(S)-2-(2-((R)-(4-(tert-Butyl)phenyl)sulfinyl)phenyl)-4-phenyl-4,5-dihydrooxazole

Precaución: for Research Use Only

Formula: C₂₅H₂₅NO₂S
CAS Number: 1965335-70-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR