4-Chloro-5-(4-((2,6-dichlorophenyl)sulfonyl)piperazin-1-yl)benzofuran-2-carboxylic acid ≥98%

Supplier: Ambeed
Danger

A1281092-50MG A1281092-100MG
77621-466EA 132.77 USD
77621-466 77867-216
4-Chloro-5-(4-((2,6-dichlorophenyl)sulfonyl)piperazin-1-yl)benzofuran-2-carboxylic acid ≥98%
4-Chloro-5-(4-((2,6-dichlorophenyl)sulfonyl)piperazin-1-yl)benzofuran-2-carboxylic acid

Precaución: for Research Use Only

Formula: C₁₉H₁₅Cl₃N₂O₅S
MDL Number: MFCD32701906
CAS Number: 1192171-69-9
UN: 3077
ADR: 9,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR