N-Acetyl-S-benzyl-L-cysteine ≥95%

Supplier: Ambeed
Warning

Synonyms: N-Ac-S-Bzl-Cys-OH

A128011-250MG A128011-1G
77599-864EA 63.55 USD
77599-864 77666-822
N-Acetyl-S-benzyl-L-cysteine ≥95%
N-Acetyl-S-benzyl-L-cysteine

Precaución: for Research Use Only

Formula: C₁₂H₁₅NO₃S
MW: 253.31 g/mol
MDL Number: MFCD00038237
CAS Number: 19542-77-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR