7-(3-Bromo-4-methoxybenzyl)-1-ethyl-8-(((1R,2R)-2-hydroxycyclopentyl)amino)-3-(2-hydroxyethyl)-3,7-dihydro-1H-purine-2,6-dione ≥98%

Supplier: Ambeed

A1280080-10MG A1280080-1MG A1280080-5MG
77596-686EA 34.85 USD
77596-686 77596-688 77596-690
7-(3-Bromo-4-methoxybenzyl)-1-ethyl-8-(((1R,2R)-2-hydroxycyclopentyl)amino)-3-(2-hydroxyethyl)-3,7-dihydro-1H-purine-2,6-dione ≥98%
7-(3-Bromo-4-methoxybenzyl)-1-ethyl-8-(((1R,2R)-2-hydroxycyclopentyl)amino)-3-(2-hydroxyethyl)-3,7-dihydro-1H-purine-2,6-dione

Precaución: for Research Use Only

Formula: C₂₂H₂₈BrN₅O₅
MDL Number: MFCD31807142
CAS Number: 569351-91-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR