2,6-Difluoro-4-(trans-4-pentylcyclohexyl)benzonitrile ≥98%

Supplier: Ambeed
Warning

A1279486-5G A1279486-100G A1279486-25G
77621-456EA 18.54 USD
77621-456 77867-201 77867-202
2,6-Difluoro-4-(trans-4-pentylcyclohexyl)benzonitrile ≥98%
2,6-Difluoro-4-(trans-4-pentylcyclohexyl)benzonitrile

Precaución: for Research Use Only

Formula: C₁₈H₂₃F₂N
CAS Number: 88308-37-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR