Fusidic acid ≥97%

Supplier: Ambeed
Warning

Synonyms: (Z)-2-((3R,4S,5S,8S,9S,10S,11R,13R,14S,16S)-16-acetoxy-3,11-dihydroxy-4,8,10,14-tetramethyldodecahydro-1H-cyclopenta[a]phenanthren-17(2H,10H,14H)-ylidene)-6-methylhept-5-enoic acid, Fusidine

A127938-100MG A127938-25G A127938-100G A127938-1G A127938-250MG A127938-5G
77596-680EA 5.13 USD
77596-680 77624-074 77876-918 77876-919 77876-920 77876-921
Fusidic acid ≥97%
Fusidic acid

Precaución: for Research Use Only

Formula: C₃₁H₄₈O₆
MDL Number: MFCD00865135
CAS Number: 6990-06-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR