1,1'-((Propane-2,2-diylbis(4,1-phenylene))bis(oxy))bis(propan-2-ol) ≥98%

Supplier: Ambeed
Warning

A1278177-100MG A1278177-250MG
77867-192EA 39 USD
77867-192 77867-193
1,1'-((Propane-2,2-diylbis(4,1-phenylene))bis(oxy))bis(propan-2-ol) ≥98%
1,1'-((Propane-2,2-diylbis(4,1-phenylene))bis(oxy))bis(propan-2-ol)

Precaución: for Research Use Only

Formula: C₂₁H₂₈O₄
MDL Number: MFCD00191826
CAS Number: 116-37-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR