trans-4-(Aminomethyl)cyclohexanecarboxylic acid ≥98%

Supplier: Ambeed
Warning

A127600-100G A127600-500G A127600-10G A127600-25G
77599-848EA 78.93 USD
77599-848 77599-850 77881-018 77881-019
trans-4-(Aminomethyl)cyclohexanecarboxylic acid ≥98%
trans-4-(Aminomethyl)cyclohexanecarboxylic acid

Precaución: for Research Use Only

Formula: C₈H₁₅NO₂
MDL Number: MFCD00001466
CAS Number: 1197-18-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR