(2,6-Dicyanophenyl)boronic acid ≥98%

Supplier: Ambeed

A1275883-100MG A1275883-1G A1275883-250MG
77867-183EA 91 USD
77867-183 77867-184 77867-185
(2,6-Dicyanophenyl)boronic acid ≥98%
(2,6-Dicyanophenyl)boronic acid

Precaución: for Research Use Only

Formula: C₈H₅BN₂O₂
MDL Number: MFCD28364141
CAS Number: 232275-35-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR