trans-3-(Cyanomethyl)-3-(4-(6-(1-methyl-1H-pyrazol-4-yl)pyrazolo[1,5-a]pyrazin-4-yl)-1H-pyrazol-1-yl)cyclobutanecarbonitrile ≥98%

Supplier: Ambeed
Warning

A1275148-10MG A1275148-50MG A1275148-5MG A1275148-100MG
77621-436EA 285.31 USD
77621-436 77621-438 77621-440 77621-434
trans-3-(Cyanomethyl)-3-(4-(6-(1-methyl-1H-pyrazol-4-yl)pyrazolo[1,5-a]pyrazin-4-yl)-1H-pyrazol-1-yl)cyclobutanecarbonitrile ≥98%
trans-3-(Cyanomethyl)-3-(4-(6-(1-methyl-1H-pyrazol-4-yl)pyrazolo[1,5-a]pyrazin-4-yl)-1H-pyrazol-1-yl)cyclobutanecarbonitrile

Precaución: for Research Use Only

Formula: C₂₀H₁₇N₉
MDL Number: MFCD32197240
CAS Number: 2127109-84-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR