(6-((5-Bromo-2-((2-methoxy-5-methyl-4-(4-(4-methylpiperazin-1-yl)piperidin-1-yl)phenyl)amino)pyrimidin-4-yl)amino)quinoxalin-5-yl)dimethylphosphine oxide ≥98%

Supplier: Ambeed
Warning

Synonyms: EGFR-IN-7

A1275056-10MG A1275056-5MG
77621-430EA 151.41 USD
77621-430 77621-432
(6-((5-Bromo-2-((2-methoxy-5-methyl-4-(4-(4-methylpiperazin-1-yl)piperidin-1-yl)phenyl)amino)pyrimidin-4-yl)amino)quinoxalin-5-yl)dimethylphosphine oxide ≥98%
(6-((5-Bromo-2-((2-methoxy-5-methyl-4-(4-(4-methylpiperazin-1-yl)piperidin-1-yl)phenyl)amino)pyrimidin-4-yl)amino)quinoxalin-5-yl)dimethylphosphine oxide

Precaución: for Research Use Only

Formula: C₃₂H₄₁BrN₉O₂P
MW: 694.6 g/mol
CAS Number: 2267329-76-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR