3-Bromo-4-chlorothieno[3,2-c]pyridine ≥98%

Supplier: Ambeed
Warning

A127424-1G A127424-5G
77670-802EA 92 USD
77670-802 77670-804
3-Bromo-4-chlorothieno[3,2-c]pyridine ≥98%
3-Bromo-4-chlorothieno[3,2-c]pyridine

Precaución: for Research Use Only

Formula: C₇H₃BrClNS
MDL Number: MFCD08275122
CAS Number: 29064-82-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR