(R)-3-Methylpiperidin-3-ol hydrochloride ≥98%

Supplier: Ambeed
Warning

A1273991-1G A1273991-250MG
77592-480EA 236.78 USD
77592-480 77596-664
(R)-3-Methylpiperidin-3-ol hydrochloride ≥98%
(R)-3-Methylpiperidin-3-ol hydrochloride

Precaución: for Research Use Only

Formula: C₆H₁₄ClNO
MW: 151.6345 g/mol
MDL Number: MFCD31693606
CAS Number: 2305080-34-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR